3,5-Dimethyl-4-(hydroxymethyl)-1-phenyl-1H-pyrazole

Lieferant: Apollo Scientific

Synonyme: (3,5-Dimethyl-1-phenyl-1H-pyrazol-4-yl)methanol

OR9983-1G
APOSOR9983-1GEA 173 EUR
APOSOR9983-1G
3,5-Dimethyl-4-(hydroxymethyl)-1-phenyl-1H-pyrazole
3,5-Dimethyl-4-(hydroxymethyl)-1-phenyl-1H-pyrazole
Formel: C₁₂H₁₄N₂O
Schmelzpunkt: 106…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06202725
CAS-Nummer: 58789-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR