5-Phenyl-1,3-oxazole
Lieferant: Apollo Scientific
|
Achtung
|
OR9992-1G
OR9992-250MG
APOSOR9992-1GEA
0
EUR
APOSOR9992-1G
APOSOR9992-250MG
5-Phenyl-1,3-oxazole
5-Phenyl-1,3-oxazole
Formel:
C9H7NO Schmelzpunkt: 40…42 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00067074 CAS-Nummer: 1006-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...