Methyl (2,3-dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-4-(2-methoxy-2-oxoethyl)-3-oxo-4H-1,4-benzothiazine, (2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetic acid methyl ester

OR9998-100MG
APOSOR9998-100MGEA 23.2 EUR
APOSOR9998-100MG
Methyl (2,3-dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetate
Methyl (2,3-dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetate
Formel: C₁₁H₁₁NO₃S
Molecular Weight: 237,275 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665898
CAS-Nummer: 702670-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR