α-Brom-2-chlor-3,6-difluortoluol 98%

Lieferant: Apollo Scientific
Gefahr

PC0008-1G PC0008-5G PC0008-25G
APOSPC0008-1GEA 23.2 EUR
APOSPC0008-1G APOSPC0008-5G APOSPC0008-25G
α-Brom-2-chlor-3,6-difluortoluol 98%
α-Brom-2-chlor-3,6-difluortoluol
Formel: C₇H₄BrClF₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631446
CAS-Nummer: 90292-67-4
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR