3-(3-(Trifluormethoxy)phenyl)propionsäure 95%

Lieferant: Apollo Scientific
Achtung

PC0014-5G PC0014-1G
APOSPC0014-5GEA 159 EUR
APOSPC0014-5G APOSPC0014-1G
3-(3-(Trifluormethoxy)phenyl)propionsäure 95%
3-(3-(Trifluormethoxy)phenyl)propionsäure
Formel: C₁₀H₉F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01317834
CAS-Nummer: 168833-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR