(2-Chlor-3,6-difluorphenyl)essigsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC0017-1G
PC0017-5G
PC0017-100MG
APOSPC0017-1GEA
116
EUR
APOSPC0017-1G
APOSPC0017-5G
APOSPC0017-100MG
(2-Chlor-3,6-difluorphenyl)essigsäure
(2-Chlor-3,6-difluorphenyl)essigsäure
Formel:
C₈H₅ClF₂O₂ Molecular Weight: 206.58 g/mol Schmelzpunkt: 136…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631448 CAS-Nummer: 261762-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...