4,4',4''-Trifluorotrityl bromide

Lieferant: Apollo Scientific

PC0030-1G
APOSPC0030-1GEA 314 EUR
APOSPC0030-1G
4,4',4''-Trifluorotrityl bromide
4,4',4''-Trifluorotrityl bromide
Formel: C₁₉H₁₂BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00191586
CAS-Nummer: 200004-38-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR