{3-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-1-yl}acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Carboxymethyl)-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole, 3-[1-(Carboxymethyl)-1H-pyrazol-3-yl]benzotrifluoride

PC0032-100MG
APOSPC0032-100MGEA 91.5 EUR
APOSPC0032-100MG
{3-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-1-yl}acetic acid
{3-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-1-yl}acetic acid
Formel: C₁₂H₉F₃N₂O₂
Molecular Weight: 270,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00973826
CAS-Nummer: 325970-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR