5-Chlor-2-fluorbenzonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC0037-100G
PC0037-500G
PC0037-50G
PC0037-25G
APOSPC0037-100GEA
97.3
EUR
APOSPC0037-100G
APOSPC0037-500G
APOSPC0037-50G
APOSPC0037-25G
5-Chlor-2-fluorbenzonitril 97%
5-Chlor-2-fluorbenzonitril
Formel:
C₇H₃ClFN Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631434 CAS-Nummer: 57381-34-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...