Boc-3-(trifluoromethyl)-D-phenylalanine
Lieferant: Apollo Scientific
PC0043-1G
APOSPC0043-1GEA
0
EUR
APOSPC0043-1G
Boc-3-(trifluoromethyl)-D-phenylalanine
Boc-3-(trifluoromethyl)-D-phenylalanine
Formel:
C₁₅H₁₈F₃NO₄ Schmelzpunkt: 135…138 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00672525 CAS-Nummer: 82317-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...