4-Vinylbenzyl trifluoroacetate

Lieferant: Apollo Scientific
Achtung

PC0052-5G
APOSPC0052-5GEA 108 EUR
APOSPC0052-5G
4-Vinylbenzyl trifluoroacetate
4-Vinylbenzyl trifluoroacetate
Formel: C₁₁H₉F₃O₂
Molecular Weight: 230.183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00080689
CAS-Nummer: 229956-99-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR