N-Methyl-2-nitro-4-(trifluormethyl)anilin
Lieferant: Apollo Scientific
|
Achtung
|
PC0054-1G
APOSPC0054-1GEA
0
EUR
APOSPC0054-1G
N-Methyl-2-nitro-4-(trifluormethyl)anilin
N-Methyl-2-nitro-4-(trifluormethyl)anilin
Formel:
C₈H₇F₃N₂O₂ Schmelzpunkt: 77…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798375 CAS-Nummer: 20200-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...