4-(Trifluormethylthio)phenylacetonitril 98%

Lieferant: Apollo Scientific
Achtung

PC0076-10G
APOSPC0076-10GEA 473 EUR
APOSPC0076-10G
4-(Trifluormethylthio)phenylacetonitril 98%
4-(Trifluormethylthio)phenylacetonitril
Formel: C₉H₆F₃NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00236354
CAS-Nummer: 70124-90-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR