6-Chloro-2-fluoro-3-methylbenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Chloro-2-fluoro-3-methylphenyl)methanol

PC0109-1G PC0109-5G PC0109-25G
APOSPC0109-1GEA 29 EUR
APOSPC0109-1G APOSPC0109-5G APOSPC0109-25G
6-Chloro-2-fluoro-3-methylbenzyl alcohol
6-Chloro-2-fluoro-3-methylbenzyl alcohol
Formel: C₈H₈ClFO
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631372
CAS-Nummer: 261762-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR