4,5-Difluoro-2-methylbenzimidazole

Lieferant: Apollo Scientific
Achtung

PC0115-100MG
APOSPC0115-100MGEA 69.8 EUR
APOSPC0115-100MG
4,5-Difluoro-2-methylbenzimidazole
4,5-Difluoro-2-methylbenzimidazole
Formel: C₈H₆F₂N₂
Molecular Weight: 168.15 g/mol
Schmelzpunkt: 250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00665871
CAS-Nummer: 229957-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR