2-Chloro-6-fluoro-3-methylcinnamic acid 97%
Lieferant: Apollo Scientific
PC0117-1G
APOSPC0117-1GEA
39.3
EUR
APOSPC0117-1G
2-Chloro-6-fluoro-3-methylcinnamic acid 97%
2-Chloro-6-fluoro-3-methylcinnamic acid
Formel:
C₁₀H₈ClFO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631396 CAS-Nummer: 261762-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...