1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate 98.5+%
Lieferant: Apollo Scientific
Synonyme:
Chloro-N,N,N',N'-bis(tetramethylene)formamidinium Tetrafluoroborate, TPyICU, 1-(Chloro(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium tetrafluoroborate
PC0125-5G
APOSPC0125-5GEA
58
EUR
APOSPC0125-5G
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate 98.5+%
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate
Formel:
C₉H₁₆BClF₄N₂ Molecular Weight: 274,49 g/mol Schmelzpunkt: 104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191334 CAS-Nummer: 115007-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...