N-Methyl-N-(trimethylsilyl)trifluoracetamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoracetamid, MSTFA
PC0132-250G
PC0132-25G
PC0132-50G
APOSPC0132-250GEA
174
EUR
APOSPC0132-250G
APOSPC0132-25G
APOSPC0132-50G
N-Methyl-N-(trimethylsilyl)trifluoracetamid 98%
N-Methyl-N-(trimethylsilyl)trifluoracetamid
A powerful silylation reagent with volatile byproducts:J. Chromat., 115, 591 (1975); 134, 387 (1977).
Formel:
C₆H₁₂F₃NOSi Siedepunkt: 130…132 °C Dichte: 1,074 g/cm³ (25 °C) Flammpunkt: 25 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00000411 CAS-Nummer: 24589-78-4 EINECS: 246-331-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...