(3-Chlor-2-fluorphenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Chloro-2-fluorophenyl)acetic acid

PC0133-1G PC0133-5G PC0133-10G
APOSPC0133-1GEA 21.9 EUR
APOSPC0133-1G APOSPC0133-5G APOSPC0133-10G
(3-Chlor-2-fluorphenyl)essigsäure
(3-Chlor-2-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Schmelzpunkt: 128…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631554
CAS-Nummer: 261762-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR