(3-Chlor-2-fluorphenyl)essigsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3-Chloro-2-fluorophenyl)acetic acid
PC0133-1G
PC0133-5G
PC0133-10G
APOSPC0133-1GEA
21.9
EUR
APOSPC0133-1G
APOSPC0133-5G
APOSPC0133-10G
(3-Chlor-2-fluorphenyl)essigsäure
(3-Chlor-2-fluorphenyl)essigsäure
Formel:
(C₆H₃ClF)CH₂COOH Schmelzpunkt: 128…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631554 CAS-Nummer: 261762-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...