(1,2-Dichlorotrifluoroethyl)cyclohexane

Lieferant: Apollo Scientific
Achtung

PC0140-5G
APOSPC0140-5GEA 348 EUR
APOSPC0140-5G
(1,2-Dichlorotrifluoroethyl)cyclohexane
(1,2-Dichlorotrifluoroethyl)cyclohexane
Formel: C₈H₁₁Cl₂F₃
Siedepunkt: 83…84 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00155754
CAS-Nummer: 219904-98-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR