3-Chlor-2-fluor-5-(trifluormethyl)anisol 98%

Lieferant: Apollo Scientific

PC0142-5G PC0142-1G PC0142-25G PC0142-100G
APOSPC0142-5GEA 0 EUR
APOSPC0142-5G APOSPC0142-1G APOSPC0142-25G APOSPC0142-100G
3-Chlor-2-fluor-5-(trifluormethyl)anisol 98%
3-Chlor-2-fluor-5-(trifluormethyl)anisol
Formel: C₈H₅ClF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631517
CAS-Nummer: 261763-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR