4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

Synonyme: 2-(4-Sulphamoylphenoxy)-3-(trifluoromethyl)pyridine

PC01471-1G
APOSPC01471-1GEA 310 EUR
APOSPC01471-1G
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formel: C₁₂H₉F₃N₂O₃S
Molecular Weight: 318,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774165
CAS-Nummer: 874839-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR