3-Fluor-2-methoxypyridin 95%

Lieferant: Apollo Scientific

PC01473-10G PC01473-25G PC01473-5G
APOSPC01473-10GEA 144 EUR
APOSPC01473-10G APOSPC01473-25G APOSPC01473-5G
3-Fluor-2-methoxypyridin 95%
3-Fluoro-2-methoxypyridine
Formel: C₆H₆FNO
Flammpunkt: <60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094945
CAS-Nummer: 884494-69-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR