3-(4-Fluorophenyl)pyrrolidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Pyrrolidin-3-yl)fluorobenzene

PC01477-1G
APOSPC01477-1GEA 669 EUR
APOSPC01477-1G
3-(4-Fluorophenyl)pyrrolidine
3-(4-Fluorophenyl)pyrrolidine
Formel: C₁₀H₁₂FN
Lagertemperatur: Raumtemperatur
MDL: MFCD03032760
CAS-Nummer: 144620-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR