1-[3-(Difluoromethoxy)phenyl]imidazolidin-2-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[3-(Difluoromethoxy)phenyl]-2-oxoimidazolidine, 1-(Difluoromethoxy)-3-(2-oxoimidazolidin-1-yl)benzene

PC01478-1G
APOSPC01478-1GEA 144 EUR
APOSPC01478-1G
1-[3-(Difluoromethoxy)phenyl]imidazolidin-2-one
1-[3-(Difluoromethoxy)phenyl]imidazolidin-2-one
Formel: C₁₀H₁₀F₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20037413
CAS-Nummer: 1378261-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR