3-Brom-5-chlorbenzotrifluorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-3-chloro-5-(trifluoromethyl)benzene

PC01480-1G
APOSPC01480-1GEA 14.7 EUR
APOSPC01480-1G
3-Brom-5-chlorbenzotrifluorid 97%
3-Brom-5-chlorbenzotrifluorid
Formel: C₇H₃BrClF₃
Molecular Weight: 259,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260894
CAS-Nummer: 928783-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR