4,7-Difluoroindan-1-one

Lieferant: Apollo Scientific

Synonyme: 4,7-Difluoro-2,3-dihydro-1H-inden-1-one, 4,7-Difluoro-2,3-dihydro-1-oxo-1H-indene

PC01482-1G
APOSPC01482-1GEA 82.8 EUR
APOSPC01482-1G
4,7-Difluoroindan-1-one
4,7-Difluoroindan-1-one
Formel: C₉H₆F₂O
Molecular Weight: 168.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772126
CAS-Nummer: 130408-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR