4,4,4-Trifluorobutane-1-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC01483-250MG
APOSPC01483-250MGEA 145 EUR
APOSPC01483-250MG
4,4,4-Trifluorobutane-1-sulphonyl chloride
4,4,4-Trifluorobutane-1-sulphonyl chloride
Formel: C₄H₆ClF₃O₂S
Molecular Weight: 210,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657959
CAS-Nummer: 212190-25-5
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR