3-Bromo-4-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC01485-1G PC01485-5G
APOSPC01485-1GEA 0 EUR
APOSPC01485-1G APOSPC01485-5G
3-Bromo-4-(trifluoromethyl)benzyl bromide 97%
3-Bromo-4-(trifluoromethyl)benzyl bromide
Formel: C₈H₅Br₂F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13185413
CAS-Nummer: 372120-77-9
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR