3-Iodo-4-(trifluoromethoxy)nitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Iodo-4-nitrophenyl trifluoromethyl ether, 2-Iodo-4-nitro-1-(trifluoromethoxy)benzene
PC01489-1G
PC01489-5G
PC01489-25G
APOSPC01489-1GEA
69.6
EUR
APOSPC01489-1G
APOSPC01489-5G
APOSPC01489-25G
3-Iodo-4-(trifluoromethoxy)nitrobenzene
3-Iodo-4-(trifluoromethoxy)nitrobenzene
Formel:
C₇H₃F₃INO₃ Molecular Weight: 333.003 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13194454 CAS-Nummer: 194344-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...