2-Nitro-4-(trifluormethyl)benzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
PC01501-1G
APOSPC01501-1GEA
87.3
EUR
APOSPC01501-1G
2-Nitro-4-(trifluormethyl)benzylalkohol
2-Nitro-4-(trifluormethyl)benzylalkohol
Formel:
C₈H₆F₃NO₃ Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095418 CAS-Nummer: 133605-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...