2-Brom-4-fluorthiophenol

Lieferant: Apollo Scientific
Achtung

PC01516-1G PC01516-250MG
APOSPC01516-1GEA 422 EUR
APOSPC01516-1G APOSPC01516-250MG
2-Brom-4-fluorthiophenol
2-Brom-4-fluorthiophenol
Formel: C₆H₄BrFS
Lagertemperatur: Kühlschrank
MDL: MFCD09475472
CAS-Nummer: 773853-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR