2-Brom-4-fluorthiophenol
Lieferant: Apollo Scientific
|
Achtung
|
PC01516-1G
PC01516-250MG
APOSPC01516-1GEA
422
EUR
APOSPC01516-1G
APOSPC01516-250MG
2-Brom-4-fluorthiophenol
2-Brom-4-fluorthiophenol
Formel:
C₆H₄BrFS Lagertemperatur: Kühlschrank |
MDL:
MFCD09475472 CAS-Nummer: 773853-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...