(3-Bromo-5-(trifluoromethyl)phenyl)methanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC01529-1G
PC01529-25G
PC01529-5G
APOSPC01529-1GEA
0
EUR
APOSPC01529-1G
APOSPC01529-25G
APOSPC01529-5G
(3-Bromo-5-(trifluoromethyl)phenyl)methanol 95%
(3-Bromo-5-(trifluoromethyl)phenyl)methanol
Formel:
C₈H₆BrF₃O Schmelzpunkt: 52…54 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835207 CAS-Nummer: 172023-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...