2-Nitrobenzenesulphonyl fluoride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Nitrobenzene-1-sulphonyl fluoride

PC01530-250MG
APOSPC01530-250MGEA 21.9 EUR
APOSPC01530-250MG
2-Nitrobenzenesulphonyl fluoride 97%
2-Nitrobenzenesulphonyl fluoride
Formel: C₆H₄FNO₄S
Schmelzpunkt: 58…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007413
CAS-Nummer: 433-98-7
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR