4-Bromo-3-fluoroisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromo-8-fluoro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Bromo-8-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione
PC01535-1G
PC01535-250MG
APOSPC01535-1GEA
580
EUR
APOSPC01535-1G
APOSPC01535-250MG
4-Bromo-3-fluoroisatoic anhydride
4-Bromo-3-fluoroisatoic anhydride
Formel:
C₈H₃BrFNO₃ Molecular Weight: 260.017 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28348374 CAS-Nummer: 1820618-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...