4-Bromo-3-fluoroisatoic anhydride

Lieferant: Apollo Scientific
Achtung

PC01535-1G PC01535-250MG
APOSPC01535-1GEA 0 EUR
APOSPC01535-1G APOSPC01535-250MG
4-Bromo-3-fluoroisatoic anhydride
4-Bromo-3-fluoroisatoic anhydride
Formel: C₈H₃BrFNO₃
Lagertemperatur: Kühlschrank
MDL: MFCD28348374
CAS-Nummer: 1820618-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR