3-Chloro-2-fluoro-5-(trifluoromethyl)benzylamine

Lieferant: Apollo Scientific

Synonyme: 3-(Aminomethyl)-5-chloro-4-fluorobenzotrifluoride, [3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl]methylamine

PC0156-1G
APOSPC0156-1GEA 424 EUR
APOSPC0156-1G
3-Chloro-2-fluoro-5-(trifluoromethyl)benzylamine
3-Chloro-2-fluoro-5-(trifluoromethyl)benzylamine
Formel: C₈H₆ClF₄N
Molecular Weight: 227,587 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631401
CAS-Nummer: 261763-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR