3-Chlor-2-fluor-5-(trifluormethyl)benzylbromid

Lieferant: Apollo Scientific
Gefahr

PC0158-1G PC0158-5G PC0158-25G
APOSPC0158-1GEA 0 EUR
APOSPC0158-1G APOSPC0158-5G APOSPC0158-25G
3-Chlor-2-fluor-5-(trifluormethyl)benzylbromid
3-Chlor-2-fluor-5-(trifluormethyl)benzylbromid
Formel: C₈H₄BrClF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01631513
CAS-Nummer: 261763-09-1
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR