1,3-Dimethoxy-1,1,2,3,3-pentafluoropropane
Lieferant: Apollo Scientific
Synonyme:
1,3-Dimethoxy-2H-perfluoropropane
PC01647-25G
PC01647-5G
APOSPC01647-25GEA
1280
EUR
APOSPC01647-25G
APOSPC01647-5G
1,3-Dimethoxy-1,1,2,3,3-pentafluoropropane
1,3-Dimethoxy-1,1,2,3,3-pentafluoropropane
Formel:
C₅H₇F₅O₂ Molecular Weight: 194.1 g/mol Siedepunkt: 64…65 °C Dichte: 1.3673 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD16170455 CAS-Nummer: 758-62-3 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...