2-Fluoro-4-methoxy-α-methylbenzyl alcohol 97%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Fluoro-4-methoxyphenyl)ethan-1-ol, 1-(2-Fluoro-4-methoxyphenyl)ethanol

PC01648-1G PC01648-250MG PC01648-5G
APOSPC01648-1GEA 290 EUR
APOSPC01648-1G APOSPC01648-250MG APOSPC01648-5G
2-Fluoro-4-methoxy-α-methylbenzyl alcohol 97%
2-Fluoro-4-methoxy-α-methylbenzyl alcohol
Formel: C₉H₁₁FO₂
Molecular Weight: 170.181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12047707
CAS-Nummer: 74457-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR