2-Fluor-4-methoxybenzylbromid

Lieferant: Apollo Scientific
Gefahr

PC01656-5G
APOSPC01656-5GEA 0 EUR
APOSPC01656-5G
2-Fluor-4-methoxybenzylbromid
2-Fluor-4-methoxybenzylbromid
Formel: C₈H₈BrFO
Lagertemperatur: Kühlschrank
CAS-Nummer: 54788-19-1
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR