(1R)-1-(3-Bromo-4-fluorophenyl)ethan-1-ol

Lieferant: Apollo Scientific
Achtung

PC01661-1G
APOSPC01661-1GEA 611 EUR
APOSPC01661-1G
(1R)-1-(3-Bromo-4-fluorophenyl)ethan-1-ol
(1R)-1-(3-Bromo-4-fluorophenyl)ethan-1-ol
Formel: C₈H₈BrFO
Lagertemperatur: Raumtemperatur
MDL: MFCD09863798
CAS-Nummer: 1212136-05-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR