5-Trifluoromethyl-1H-benzoimidazole-2-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

PC01663-1G
APOSPC01663-1GEA 473 EUR
APOSPC01663-1G
5-Trifluoromethyl-1H-benzoimidazole-2-carboxylic acid 95%
5-Trifluoromethyl-1H-benzoimidazole-2-carboxylic acid
Formel: C₉H₅F₃N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD11052827
CAS-Nummer: 2107-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR