4-Chloro-2-(trifluoromethyl)pyridine 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC01677-25G
PC01677-5G
PC01677-1G
APOSPC01677-25GEA
0
EUR
APOSPC01677-25G
APOSPC01677-5G
APOSPC01677-1G
4-Chloro-2-(trifluoromethyl)pyridine 99%
4-Chloro-2-(trifluoromethyl)pyridine
Formel:
C₆H₃ClF₃N Siedepunkt: 137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368047 CAS-Nummer: 131748-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...