1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulphonyl)imide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-MEthyl-1-propylpyrrolidinium bis(trifluoromethanesulphonyl)imide

PC01683-25G
APOSPC01683-25GEA 95.7 EUR
APOSPC01683-25G
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulphonyl)imide 98%
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulphonyl)imide
Formel: C₁₀H₁₈F₆N₂O₄S₂
Molecular Weight: 408.37 g/mol
Schmelzpunkt: 12 °C
Dichte: 1.44 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD08458917
CAS-Nummer: 223437-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR