3-Fluoro-4-phenoxybenzoic acid 95%

Lieferant: Apollo Scientific

PC01685-250MG
APOSPC01685-250MGEA 284 EUR
APOSPC01685-250MG
3-Fluoro-4-phenoxybenzoic acid 95%
3-Fluoro-4-phenoxybenzoic acid
Formel: C₁₃H₉FO₃
Molecular Weight: 232,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11156619
CAS-Nummer: 129623-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR