3-Chloro-4-(trifluoromethoxy)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC0174-1G PC0174-25G PC0174-5G
APOSPC0174-1GEA 0 EUR
APOSPC0174-1G APOSPC0174-25G APOSPC0174-5G
3-Chloro-4-(trifluoromethoxy)benzaldehyde
3-Chloro-4-(trifluoromethoxy)benzaldehyde
Formel: C8H4ClF3O2
Lagertemperatur: Raumtemperatur
MDL: MFCD01631561
CAS-Nummer: 83279-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR