3-Chlor-4-(trifluormethoxy)benzonitril
Lieferant: Apollo Scientific
|
Achtung
|
PC0178-1G
PC0178-5G
PC0178-10G
APOSPC0178-1GEA
21.9
EUR
APOSPC0178-1G
APOSPC0178-5G
APOSPC0178-10G
3-Chlor-4-(trifluormethoxy)benzonitril
3-Chlor-4-(trifluormethoxy)benzonitril
Formel:
C₈H₃ClF₃NO Molecular Weight: 221.57 g/mol Schmelzpunkt: 39…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631557 CAS-Nummer: 129604-26-8 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...